Assay Details
| Gene Symbol: Fam229b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256519.2 | XP_006256581.1 | 1 - 2 | 96 | 79 | |
| XM_008772971.1 | XP_008771193.1 | 1 - 2 | 95 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006256519.2, XM_008772971.1 transcripts.
Set Membership: |
|

Fam229b
Genomic Map