Assay Details
| Gene Symbol: Fam229b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134695.1 | NP_001128167.1 | 2 - 3 | 160 | 99 | |
| XM_008772972.1 | XP_008771194.1 | 2 - 3 | 228 | 99 | ||
| XM_008772973.1 | XP_008771195.1 | 2 - 3 | 152 | 99 | ||
| GenBank mRNA | BC158777.1 | - | 2 - 3 | 160 | 7133926 | 99 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC158777.1, NM_001134695.1, XM_008772972.1, XM_008772973.1 transcripts.
Set Membership: |
|

Fam229b
Genomic Map