Assay Details
| Gene Symbol: Fam229b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134695.1 | NP_001128167.1 | 3 - 4 | 312 | 118 | |
| XM_008772971.1 | XP_008771193.1 | 3 - 4 | 378 | 118 | ||
| XM_008772972.1 | XP_008771194.1 | 3 - 4 | 380 | 118 | ||
| XM_008772973.1 | XP_008771195.1 | 3 - 4 | 304 | 118 | ||
| GenBank mRNA | BC158777.1 | - | 3 - 4 | 312 | 7133926 | 118 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC158777.1, NM_001134695.1, XM_008772971.1, XM_008772972.1, XM_008772973.1 transcripts.
Set Membership: |
|

Fam229b
Genomic Map