Assay Details
| Gene Symbol: RGD1562114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109283.1 | NP_001102753.1 | 2 - 3 | 361 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562114| Gene Symbol: RGD1562114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109283.1 | NP_001102753.1 | 2 - 3 | 361 | 78 | |
Set Membership: |
|