Assay Details
| Gene Symbol: Crnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761263.1 | XP_008759485.1 | 2 - 3 | 177 | 82 | |
| XM_227367.5 | XP_227367.5 | 1 - 2 | 220 | 82 | ||
More Information
Set Membership: |
|

Crnn
Genomic Map