Assay Details
| Gene Symbol: Wipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147211.1 | NP_671744.1 | 7 - 8 | 1524 | 95 | |
| XM_008762877.1 | XP_008761099.1 | 8 - 9 | 2059 | 95 | ||
| GenBank mRNA | U25281.1 | - | 7 - 8 | 1524 | 95 | |
| U31159.1 | - | 8 - 9 | 1626 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P278387 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P163416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112754 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P163416 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P278387 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P163416 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P278387 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P278387 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P163416 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P174181 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P278387 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702152919 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6071 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51375 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13400 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91189 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35382 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70028 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 702152919 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10855641 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10855649 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10855638 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10855640 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10855643 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10855647 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10855645 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10855639 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10855642 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10855644 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10855648 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10855646 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10855637 | - | - |
| Rat Genome 230 2.0 Array | 1370648_a_at | - | - |
| Rat Genome 230 2.0 Array | 1370596_a_at | - | - |
| Rat Genome 230 2.0 Array | 1370648_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1370596_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1370596_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1370648_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1370648_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1370596_a_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400006792.rn.1 | TC0400000952.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400006791.rn.1 | TC0400000952.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370596_PM_a_at | - | - |
| Rat Expression Set 230 | 1370596_a_at | - | - |
| Rat Expression Set 230 | 1370648_PM_a_at | - | - |
| Rat Expression Set 230 | 1370648_a_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10855637 | - | - |
| Rat Genome U34 Set | U31160mRNA_s_at | - | - |
| Rat Genome U34 Set | U31159_s_at | - | - |
| Rat Genome U34 Set | U25281_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21270375 | - | - |

Wipf3
Genomic Map