Assay Details
Gene Symbol: Wipf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147211.1 | NP_671744.1 | 3 - 4 | 413 | 113 | |
XM_008762874.1 | XP_008761096.1 | 3 - 4 | 848 | 113 | ||
XM_008762875.1 | XP_008761097.1 | 3 - 4 | 3175 | 113 | ||
XM_008762876.1 | XP_008761098.1 | 3 - 4 | 848 | 113 | ||
XM_008762877.1 | XP_008761099.1 | 3 - 4 | 846 | 113 | ||
GenBank mRNA | U25281.1 | - | 3 - 4 | 413 | 113 | |
U31159.1 | - | 3 - 4 | 413 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
