Assay Details
| Gene Symbol: Mxi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013160.2 | NP_037292.2 | 4 - 5 | 391 | 70 | |
| XM_006231604.1 | XP_006231666.1 | 4 - 5 | 950 | 70 | ||
| XM_006231605.1 | XP_006231667.1 | 4 - 5 | 362 | 70 | ||
| XM_006231606.1 | XP_006231668.1 | 4 - 5 | 521 | 70 | ||
| GenBank mRNA | AF003008.1 | - | 4 - 5 | 344 | 70 | |
| BC081824.1 | - | 4 - 5 | 391 | 7133853 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mxi1
Genomic Map