Assay Details
| Gene Symbol: Peo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107599.1 | NP_001101069.1 | 1 - 2 | 1955 | 114 | |
| XM_008760429.1 | XP_008758651.1 | 1 - 2 | 2015 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Peo1
Genomic Map