Assay Details
| Gene Symbol: Sh3pxd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107606.1 | NP_001101076.1 | 13 - 14 | 1353 | 66 | |
| XM_006231513.2 | XP_006231575.1 | 14 - 15 | 1770 | 66 | ||
| XM_006231514.2 | XP_006231576.1 | 13 - 14 | 1690 | 66 | ||
| XM_008760439.1 | XP_008758661.1 | 13 - 14 | 1725 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P000981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97021 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000981 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000981 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P000981 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P23290 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P412452 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45562 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13222 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23475 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14240 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10730673 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730665 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730662 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730671 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730667 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730666 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730669 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730664 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730663 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730672 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730668 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730675 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730670 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730674 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10730661 | - | - |
| Rat Genome 230 2.0 Array | 1371485_at | - | - |
| Rat Genome 230 2.0 Array | 1371485_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100063358.rn.1 | TC0100007847.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10730661 | - | - |
| Rat Genome U34 Set | rc_AI138069_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20932126 | - | - |
| ABI Rat Genome Survey Microarray | 22401408 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650731 | - | - |

Sh3pxd2a
Genomic Map