Assay Details
| Gene Symbol: Erlin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106353.1 | NP_001099823.1 | 11 - 12 | 929 | 110 | |
| XM_006231441.2 | XP_006231503.1 | 11 - 12 | 949 | 110 | ||
| GenBank mRNA | BC161938.1 | - | 11 - 12 | 881 | 9022977 | 110 |
| FQ209728.1 | - | 11 - 12 | 950 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P468618 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P726392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56082 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P468618 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P726392 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P468618 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P726392 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P468618 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P726392 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P468618 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 48204 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103917 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10715525 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715528 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715520 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715527 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715526 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715522 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715524 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715521 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715530 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715523 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715529 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10715519 | - | - |
| Rat Genome 230 2.0 Array | 1371580_at | - | - |
| Rat Genome 230 2.0 Array | 1371580_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371580_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371580_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100062459.rn.1 | TC0100007758.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100062460.rn.1 | TC0100007758.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100062458.rn.1 | TC0100007758.rn.1 | RTA TC |
| Rat Expression Set 230 | 1371580_PM_at | - | - |
| Rat Expression Set 230 | 1371580_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715519 | - | - |
| Rat Genome U34 Set | rc_AI105196_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22165676 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356408 | - | - |

Erlin1
Genomic Map