Assay Details
| Gene Symbol: Cfap126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109075.1 | NP_001102545.1 | 2 - 3 | 338 | 67 | |
| XM_008769751.1 | XP_008767973.1 | 1 - 2 | 945 | 67 | ||
| XM_008769752.1 | XP_008767974.1 | 1 - 2 | 945 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109075.1, XM_008769751.1, XM_008769752.1 transcripts.
Set Membership: |
|

Cfap126
Genomic Map