Assay Details
| Gene Symbol: Cfap126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109075.1 | NP_001102545.1 | 4 - 5 | 485 | 121 | |
| XM_008769751.1 | XP_008767973.1 | 3 - 4 | 1092 | 121 | ||
| GenBank mRNA | FQ219903.1 | - | 4 - 5 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cfap126
Genomic Map