Assay Details
| Gene Symbol: Osbpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013079.1 | NP_001013097.1 | 13 - 14 | 1512 | 88 | |
| XM_006235816.2 | XP_006235878.1 | 14 - 15 | 1761 | 88 | ||
| GenBank mRNA | BC091208.1 | - | 13 - 14 | 1512 | 7381353 | 88 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P619008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81608 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P619008 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P619008 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P619008 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P194894 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34501 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24556 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86512 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29984 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10842793 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842784 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842788 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842785 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842787 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842789 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842782 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842792 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842786 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842790 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842794 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842791 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842783 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10842781 | - | - |
| Rat Genome 230 2.0 Array | 1393873_s_at | - | - |
| Rat Genome 230 2.0 Array | 1385582_at | - | - |
| Rat Genome 230 2.0 Array | 1385582_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1367519_at | - | - |
| Rat Genome 230 2.0 Array | 1367519_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393873_PM_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1367519_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385582_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1367519_at | - | - |
| Affymetrix HT Rat Focus Array | 1385582_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300039351.rn.1 | TC0300005088.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393873_PM_s_at | - | - |
| Rat Expression Set 230 | 1393873_s_at | - | - |
| Rat Expression Set 230 | 1367519_PM_at | - | - |
| Rat Expression Set 230 | 1367519_at | - | - |
| Rat Expression Set 230 | 1385582_at | - | - |
| Rat Expression Set 230 | 1367519_PM_at | - | - |
| Rat Expression Set 230 | 1385582_PM_at | - | - |
| Rat Expression Set 230 | 1367519_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842781 | - | - |
| Rat Genome U34 Set | rc_AI639528_g_at | - | - |
| Rat Genome U34 Set | rc_AI639528_at | - | - |
| Rat Genome U34 Set | rc_AI013946_at | - | - |
| Rat Genome U34 Set | rc_AI177447_at | - | - |
| Rat Genome U34 Set | rc_AI070596_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20781350 | - | - |

Osbpl2
Genomic Map