Assay Details
| Gene Symbol: Rxfp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012475.1 | NP_001012493.1 | 17 - 18 | 1956 | 138 | |
| GenBank mRNA | AY906861.1 | - | 17 - 18 | 1956 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
oocyte maturation
adenylate cyclase-activating G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
negative regulation of cell proliferation
male gonad development
positive regulation of cAMP biosynthetic process
negative regulation of apoptotic process
protein-hormone receptor activity
peptide hormone binding
oocyte maturation
adenylate cyclase-activating G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
negative regulation of cell proliferation
male gonad development
positive regulation of cAMP biosynthetic process
negative regulation of apoptotic process
protein-hormone receptor activity
peptide hormone binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P154138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13069 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P154138 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P154138 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P154138 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P154138 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28721 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19913 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 60105 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4665 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74264 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21978 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10759929 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759924 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759936 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759935 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759938 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759927 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759932 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759931 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759940 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759926 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759939 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759928 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759934 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759937 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759933 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759925 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10759930 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10759923 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1200008517.rn.1 | TC1200000975.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10759923 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20753214 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371468 | - | - |

Rxfp2
Genomic Map