Assay Details
Gene Symbol: Epha5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169137.2 | NP_001162608.1 | 1 - 2 | 538 | 63 | |
NM_024367.1 | NP_077343.1 | 1 - 2 | 538 | 63 | ||
XM_006250818.2 | XP_006250880.1 | 1 - 2 | 1047 | 63 | ||
XM_006250819.2 | XP_006250881.1 | 1 - 2 | 1047 | 63 | ||
XM_008770059.1 | XP_008768281.1 | 1 - 2 | 1047 | 63 | ||
GenBank mRNA | X78689.1 | - | 1 - 2 | 605 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
