Assay Details
| Gene Symbol: Paics | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080910.3 | NP_543186.1 | 1 - 2 | 50 | 53 | |
| GenBank mRNA | BC085711.1 | - | 1 - 2 | 50 | 7192781 | 53 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC085711.1, NM_080910.3 transcripts.
Set Membership: |
|

Paics
Genomic Map