Assay Details
| Gene Symbol: Art3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012034.1 | NP_001012034.1 | 8 - 9 | 1057 | 76 | |
| XM_006250727.1 | XP_006250789.1 | 8 - 9 | 1136 | 76 | ||
| XM_006250728.1 | XP_006250790.1 | 7 - 8 | 1106 | 76 | ||
| XM_008770026.1 | XP_008768248.1 | 8 - 9 | 1080 | 76 | ||
| XM_008770027.1 | XP_008768249.1 | 8 - 9 | 1112 | 76 | ||
| GenBank mRNA | BC079018.1 | - | 8 - 9 | 1057 | 7113746 | 76 |
| FQ214932.1 | - | 8 - 9 | 999 | 76 | ||
| FQ216328.1 | - | 8 - 9 | 1080 | 76 | ||
| FQ224878.1 | - | 8 - 9 | 1068 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P14648 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P151226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168937 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14648 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P151226 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14648 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P151226 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P151226 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P14648 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P240881 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26359 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70905 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104100 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85304 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65200 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65801 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46121 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37646 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33348 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10775807 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775815 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775816 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775813 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775810 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775806 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775809 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775814 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775811 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775820 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775808 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10775805 | - | - |
| Rat Genome 230 2.0 Array | 1372065_at | - | - |
| Rat Genome 230 2.0 Array | 1372065_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372065_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372065_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400010887.rn.1 | TC1400001483.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372065_PM_at | - | - |
| Rat Expression Set 230 | 1372065_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775805 | - | - |
| Rat Genome U34 Set | rc_AI104393_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21918128 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375337 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376636 | - | - |

Art3
Genomic Map