Assay Details
| Gene Symbol: Rufy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025127.1 | NP_001020298.1 | 3 - 4 | 805 | 75 | |
| XM_006250781.2 | XP_006250843.1 | 3 - 4 | 1669 | 75 | ||
| XM_006250782.2 | XP_006250844.1 | 3 - 4 | 1878 | 75 | ||
| XM_006250783.2 | XP_006250845.1 | 3 - 4 | 1668 | 75 | ||
| XM_006250784.2 | XP_006250846.1 | 2 - 3 | 394 | 75 | ||
| XM_006250785.2 | XP_006250847.1 | 2 - 3 | 588 | 75 | ||
| XM_008770044.1 | XP_008768266.1 | 2 - 3 | 606 | 75 | ||
| XM_008770045.1 | XP_008768267.1 | 2 - 3 | 606 | 75 | ||
| XM_008770046.1 | XP_008768268.1 | 3 - 4 | 1863 | 75 | ||
| XM_008770047.1 | XP_008768269.1 | 2 - 3 | 606 | 75 | ||
| XM_008770048.1 | XP_008768270.1 | 4 - 5 | 1722 | 75 | ||
| GenBank mRNA | BC089952.1 | - | 3 - 4 | 805 | 7312240 | 75 |
| EF538802.1 | - | 3 - 4 | 410 | 75 | ||
| EF577045.1 | - | 2 - 3 | 356 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rufy3
Genomic Map