Assay Details
| Gene Symbol: Btc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022256.2 | NP_071592.1 | 215 | 112 | ||
| GenBank mRNA | AB028862.1 | - | 215 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Btc
Genomic Map