Assay Details
| Gene Symbol: Ndc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025023.1 | NP_001020194.1 | 11 - 12 | 1564 | 66 | |
| GenBank mRNA | BC079443.1 | - | 11 - 12 | 1564 | 7115485 | 66 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P652332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59521 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P652332 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P652332 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P652332 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P652332 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59445 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37024 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10870685 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870681 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870680 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870694 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870689 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870683 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870688 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870690 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870692 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870686 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870684 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870679 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870687 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870682 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870691 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870693 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10870695 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10870678 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500007620.rn.1 | TC0500001143.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500007626.rn.1 | TC0500001143.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500007622.rn.1 | TC0500001143.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10870678 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368592 | - | - |

Ndc1
Genomic Map