Assay Details
| Gene Symbol: Fam151a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005558.1 | NP_001005558.1 | 3 - 4 | 462 | 62 | |
| GenBank mRNA | BC081977.1 | - | 3 - 4 | 462 | 7127557 | 62 |
More Information
Updated Mapping Information
Set Membership: |
|

Fam151a
Genomic Map