Assay Details
| Gene Symbol: Foxj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008764012.1 | 12 - 13 | 2048 | 66 | ||
| XM_008764013.1 | XP_008762235.1 | 10 - 11 | 3971 | 66 | ||
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P068648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60536 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P068648 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P068648 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P068648 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P22256 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47006 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101372 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 56037 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10871599 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871598 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871607 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871600 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871604 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871605 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871602 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871609 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871608 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871601 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871603 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871606 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10871597 | - | - |
| Rat Genome 230 2.0 Array | 1377347_at | - | - |
| Rat Genome 230 2.0 Array | 1377347_PM_at | - | - |
| Rat Expression Set 230 | 1377347_at | - | - |
| Rat Expression Set 230 | 1377347_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871597 | - | - |
| Rat Genome U34 Set | rc_AA925959_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22327603 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1348889 | - | - |

Foxj3
Genomic Map