Assay Details
| Gene Symbol: Neo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766432.1 | XP_008764654.1 | 3 - 4 | 833 | 64 | |
| GenBank mRNA | U68726.1 | - | 791 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Neo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766432.1 | XP_008764654.1 | 3 - 4 | 833 | 64 | |
| GenBank mRNA | U68726.1 | - | 791 | 64 | ||
Set Membership: |
|