Assay Details
| Gene Symbol: Cyp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012540.2 | NP_036672.2 | 4 - 5 | 1123 | 76 | |
| XM_006243150.2 | XP_006243212.1 | 4 - 5 | 1120 | 76 | ||
| GenBank mRNA | X00469.1 | - | 4 - 5 | 1123 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp1a1
Genomic Map