Assay Details
| Gene Symbol: Layn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191997.1 | NP_001178926.1 | 5 - 6 | 600 | 100 | |
| XM_006243049.2 | XP_006243111.1 | 4 - 5 | 1565 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Layn
Genomic Map