Assay Details
| Gene Symbol: Asb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037367.1 | NP_001032444.1 | 3 - 4 | 1062 | 100 | |
| XM_006257044.2 | XP_006257106.1 | 2 - 3 | 1015 | 100 | ||
| GenBank mRNA | BC107911.1 | - | 3 - 4 | 1062 | 7453889 | 100 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1026984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82099 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1026984 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1026984 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1026984 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1026984 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13502 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76183 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3639 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8999 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79368 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10938492 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938489 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938490 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938491 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10938488 | - | - |
| Rat Genome 230 2.0 Array | 1374989_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374989_at | - | - |
| Affymetrix HT Rat Focus Array | 1374989_at | - | - |
| Affymetrix HT Rat Focus Array | 1374989_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0X00011964.rn.1 | TC0X00001889.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00011967.rn.1 | TC0X00001889.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00011959.rn.1 | TC0X00001889.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00011961.rn.1 | TC0X00001889.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00011968.rn.1 | TC0X00001889.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00011962.rn.1 | TC0X00001889.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00011963.rn.1 | TC0X00001889.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374989_at | - | - |
| Rat Expression Set 230 | 1374989_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938488 | - | - |
| Rat Genome U34 Set | rc_AA925141_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20824209 | - | - |

Asb12
Genomic Map