Assay Details
| Gene Symbol: RGD1311847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013879.1 | NP_001013901.1 | 2 - 3 | 790 | 61 | |
| XM_006252799.2 | XP_006252861.1 | 2 - 3 | 847 | 61 | ||
| XM_008771073.1 | XP_008769295.1 | 2 - 3 | 840 | 61 | ||
| GenBank mRNA | BC079430.1 | - | 2 - 3 | 790 | 7114707 | 61 |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1311847
Genomic Map