Assay Details
| Gene Symbol: RGD1311847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013879.1 | NP_001013901.1 | 5 - 6 | 1521 | 66 | |
| XM_006252799.2 | XP_006252861.1 | 5 - 6 | 1578 | 66 | ||
| GenBank mRNA | BC079430.1 | - | 5 - 6 | 1521 | 7114707 | 66 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RGD1311847
Genomic Map