Assay Details
| Gene Symbol: Abtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005902.1 | NP_001005902.1 | 3 - 4 | 241 | 66 | |
| GenBank mRNA | BC083579.1 | - | 3 - 4 | 241 | 7128126 | 66 |
More Information
Updated Mapping Information
Set Membership: |
|

Abtb1
Genomic Map