Assay Details
| Gene Symbol: Ttc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004246.2 | NP_001004246.1 | 17 - 18 | 1629 | 54 | |
| XM_006243004.2 | XP_006243066.1 | 18 - 19 | 2442 | 54 | ||
| XM_006243005.2 | XP_006243067.1 | 18 - 19 | 1697 | 54 | ||
| XM_008766196.1 | XP_008764418.1 | 18 - 19 | 1844 | 54 | ||
| GenBank mRNA | BC079409.1 | - | 17 - 18 | 1631 | 7113087 | 54 |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc12
Genomic Map