Assay Details
| Gene Symbol: Dixdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037654.1 | NP_001032743.1 | 17 - 18 | 1787 | 67 | |
| XM_006243042.2 | XP_006243104.1 | 18 - 19 | 2340 | 67 | ||
| XM_006243045.2 | XP_006243107.1 | 17 - 18 | 1695 | 67 | ||
| XM_008766295.1 | XP_008764517.1 | 18 - 19 | 2376 | 67 | ||
| XM_008766296.1 | XP_008764518.1 | 14 - 15 | 1707 | 67 | ||
| GenBank mRNA | AY770507.1 | - | 17 - 18 | 1787 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cerebellar cortex development
cerebral cortex cell migration
cerebral cortex radially oriented cell migration
cell proliferation in forebrain
forebrain ventricular zone progenitor cell division
positive regulation of Wnt signaling pathway
regulation of actin cytoskeleton organization
negative regulation of neuron differentiation
positive regulation of JNK cascade
positive regulation of axonogenesis
canonical Wnt signaling pathway
regulation of microtubule cytoskeleton organization
positive regulation of canonical Wnt signaling pathway
actin binding
protein domain specific binding
mitogen-activated protein kinase kinase kinase binding
gamma-tubulin binding
cerebellar cortex development
cerebral cortex cell migration
cerebral cortex radially oriented cell migration
cell proliferation in forebrain
forebrain ventricular zone progenitor cell division
positive regulation of Wnt signaling pathway
regulation of actin cytoskeleton organization
negative regulation of neuron differentiation
positive regulation of JNK cascade
positive regulation of axonogenesis
canonical Wnt signaling pathway
regulation of microtubule cytoskeleton organization
positive regulation of canonical Wnt signaling pathway
actin binding
protein domain specific binding
mitogen-activated protein kinase kinase kinase binding
gamma-tubulin binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P454311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704171 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35724 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P454311 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P454311 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P454311 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P326742 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21225 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50391 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20824 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18812 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85733 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63346 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10917278 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917261 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917264 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917272 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917265 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917277 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917270 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917266 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917282 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917281 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917268 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917279 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917271 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917262 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917275 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917263 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917269 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10917260 | - | - |
| Rat Genome 230 2.0 Array | 1390127_at | - | - |
| Rat Genome 230 2.0 Array | 1390127_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390127_at | - | - |
| Affymetrix HT Rat Focus Array | 1390127_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800020257.rn.1 | TC0800002517.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800020261.rn.1 | TC0800002517.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800020256.rn.1 | TC0800002517.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800020260.rn.1 | TC0800002517.rn.1 | RTA TC |
| Rat Expression Set 230 | 1390127_at | - | - |
| Rat Expression Set 230 | 1390127_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917260 | - | - |
| Rat Genome U34 Set | rc_AA924354_at | - | - |
| Rat Genome U34 Set | rc_AI233394_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21242581 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370741 | - | - |

Dixdc1
Genomic Map