Assay Details
| Gene Symbol: Cse1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108607.1 | NP_001102077.1 | 23 - 24 | 2685 | 64 | |
| XM_006235648.2 | XP_006235710.1 | 24 - 25 | 2783 | 64 | ||
| GenBank mRNA | FQ233199.1 | - | 4 - 5 | 358 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cse1l
Genomic Map