Assay Details
| Gene Symbol: Mageb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109367.1 | NP_001102837.1 | 1 | 949 | 94 | |
| XM_006256984.2 | XP_006257046.1 | 2 | 1165 | 94 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mageb7
Genomic Map