Assay Details
| Gene Symbol: LOC100911685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750763.3 | XP_003750811.1 | 1564 | 58 | ||
| XM_006245740.2 | XP_006245802.1 | 1567 | 58 | |||
| Gene Symbol: Gspt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003978.1 | NP_001003978.1 | 7 - 8 | 1141 | 58 | |
| GenBank mRNA | BC079092.1 | - | 7 - 8 | 1141 | 7123818 | 58 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911685
Genomic Map