Assay Details
| Gene Symbol: Gkn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039686.1 | NP_001034775.1 | 4 - 5 | 347 | 108 | |
| XM_008763064.1 | XP_008761286.1 | 3 - 4 | 1875 | 108 | ||
| XM_008763065.1 | XP_008761287.1 | 7 - 8 | 831 | 108 | ||
| GenBank mRNA | AY943907.1 | - | 4 - 5 | 347 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gkn2
Genomic Map