Assay Details
| Gene Symbol: Prrt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749809.3 | XP_003749857.1 | 3 - 4 | 1456 | 166 | |
| XM_006237062.2 | XP_006237124.1 | 3 - 4 | 1534 | 166 | ||
| XM_006237064.2 | XP_006237126.1 | 3 - 4 | 1379 | 166 | ||
| XM_008763200.1 | XP_008761422.1 | 2 - 3 | 1378 | 166 | ||
More Information
Set Membership: |
|

Prrt3
Genomic Map