Assay Details
| Gene Symbol: Serpinb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130404.1 | NP_569088.1 | 2 - 3 | 214 | 128 | |
| XM_006249629.1 | XP_006249691.1 | 3 - 4 | 294 | 128 | ||
| XM_008769420.1 | XP_008767642.1 | 3 - 4 | 270 | 128 | ||
| GenBank mRNA | AF105329.1 | - | 2 - 3 | 214 | 128 | |
| FQ228843.1 | - | 3 - 4 | 276 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serpinb7
Genomic Map