Assay Details
| Gene Symbol: Sh3bp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022693.4 | NP_073184.3 | 2 - 3 | 257 | 62 | |
| GenBank mRNA | FM034920.1 | - | 2 - 3 | 257 | 62 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_022693.4 transcripts.
Set Membership: |
|

Sh3bp4
Genomic Map