Assay Details
| Gene Symbol: Cinp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106758.2 | NP_001100228.1 | 3 - 4 | 343 | 71 | |
| GenBank mRNA | BC158883.1 | - | 3 - 4 | 343 | 7937483 | 71 |
More Information
Updated Mapping Information
Set Membership: |
|

Cinp
Genomic Map