Assay Details
Gene Symbol: Usp9x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135893.1 | NP_001129365.1 | 4 - 5 | 743 | 68 | |
NM_001135923.1 | NP_001129395.1 | 3 - 4 | 616 | 68 | ||
XM_006256666.2 | XP_006256728.1 | 6 - 7 | 1000 | 68 | ||
XM_006256667.2 | XP_006256729.1 | 7 - 8 | 1008 | 68 | ||
XM_006256668.2 | XP_006256730.1 | 6 - 7 | 993 | 68 | ||
XM_006256669.2 | XP_006256731.1 | 3 - 4 | 639 | 68 | ||
XM_006256670.2 | XP_006256732.1 | 6 - 7 | 1259 | 68 | ||
XM_006256671.2 | XP_006256733.1 | 3 - 4 | 640 | 68 | ||
XM_008773061.1 | XP_008771283.1 | 6 - 7 | 1259 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
