Assay Details
| Gene Symbol: Acsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772664.1 | XP_008770886.1 | 3 - 4 | 883 | 65 | |
| XM_008772665.1 | XP_008770887.1 | 2 - 3 | 350 | 65 | ||
| XM_574249.6 | XP_574249.4 | 3 - 4 | 1040 | 65 | ||
More Information
Set Membership: |
|

Acsf3
Genomic Map