Assay Details
| Gene Symbol: Osgin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138504.2 | NP_612513.1 | 1 - 2 | 298 | 128 | |
| GenBank mRNA | AF549441.1 | - | 1 - 2 | 298 | 128 | |
| AF549442.1 | - | 1 - 2 | 298 | 128 | ||
| BC091111.1 | - | 1 - 2 | 294 | 7106386 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF549441.1, AF549442.1, BC091111.1, NM_138504.2 transcripts.
Set Membership: |
|

Osgin1
Genomic Map