Assay Details
| Gene Symbol: Osgin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138504.2 | NP_612513.1 | 2 - 3 | 397 | 111 | |
| XM_006255706.2 | XP_006255768.1 | 5 - 6 | 456 | 111 | ||
| GenBank mRNA | AF549441.1 | - | 2 - 3 | 397 | 111 | |
| BC091111.1 | - | 2 - 3 | 393 | 7106386 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF549441.1, BC091111.1, NM_138504.2, XM_006255706.2 transcripts.
Set Membership: |
|

Osgin1
Genomic Map