Assay Details
| Gene Symbol: NEWGENE_1305243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_341690.7 | XP_341691.7 | 638 | 70 | ||
| GenBank mRNA | FQ230829.1 | - | 590 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEWGENE_1305243| Gene Symbol: NEWGENE_1305243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_341690.7 | XP_341691.7 | 638 | 70 | ||
| GenBank mRNA | FQ230829.1 | - | 590 | 70 | ||
Set Membership: |
|