Assay Details
| Gene Symbol: Hsd17b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024391.1 | NP_077367.1 | 1 - 2 | 315 | 134 | |
| GenBank mRNA | BC088134.1 | - | 1 - 2 | 306 | 7326035 | 134 |
| X91234.1 | - | 1 - 2 | 315 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b2
Genomic Map