Assay Details
| Gene Symbol: Cdh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255559.2 | XP_006255621.1 | 15 - 16 | 2818 | 63 | |
| GenBank mRNA | AF177683.1 | - | 1 - 2 | 149 | 63 | |
More Information
Set Membership: |
|

Cdh3
Genomic Map