Assay Details
| Gene Symbol: Efcab7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191866.1 | NP_001178795.1 | 10 - 11 | 1439 | 103 | |
| XM_008763885.1 | XP_008762107.1 | 10 - 11 | 1489 | 103 | ||
| XM_008763886.1 | XP_008762108.1 | 10 - 11 | 1427 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Efcab7
Genomic Map