Assay Details
| Gene Symbol: Inadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238423.2 | XP_006238485.1 | 39 - 40 | 5184 | 61 | |
| XM_006238424.2 | XP_006238486.1 | 39 - 40 | 5190 | 61 | ||
| XM_006238425.2 | XP_006238487.1 | 14 - 15 | 2226 | 61 | ||
| XM_008763916.1 | XP_008762138.1 | 15 - 16 | 2280 | 61 | ||
More Information
Set Membership: |
|

Inadl
Genomic Map