Assay Details
| Gene Symbol: Adamts14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107636.1 | NP_001101106.1 | 13 - 14 | 2053 | 59 | |
| XM_006256449.2 | XP_006256511.1 | 14 - 15 | 2319 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adamts14
Genomic Map